Assay Details
| Gene Symbol: AT5G48570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124233.3 | NP_199668.1 | 8 - 9 | 1365 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G48570| Gene Symbol: AT5G48570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124233.3 | NP_199668.1 | 8 - 9 | 1365 | 80 | |
Set Membership: |
|