Assay Details
| Gene Symbol: FKBP15-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124234.4 | NP_199669.1 | 5 - 6 | 738 | 81 | |
| GenBank mRNA | AF370159.1 | - | 4 - 5 | 296 | 81 | |
| AY059146.1 | - | 4 - 5 | 278 | 81 | ||
| AY085294.1 | - | 4 - 5 | 299 | 81 | ||
More Information
Set Membership: |
|

FKBP15-2
Genomic Map