Assay Details
| Gene Symbol: FKBP15-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124234.4 | NP_199669.1 | 6 - 7 | 838 | 118 | |
| GenBank mRNA | AF370159.1 | - | 5 - 6 | 396 | 118 | |
| AY059146.1 | - | 5 - 6 | 378 | 118 | ||
| AY085294.1 | - | 5 - 6 | 399 | 118 | ||
More Information
Set Membership: |
|

FKBP15-2
Genomic Map