Assay Details
| Gene Symbol: AT5G48650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124241.4 | NP_199676.2 | 4 - 5 | 704 | 115 | |
| GenBank mRNA | BT002864.1 | - | 4 - 5 | 642 | 115 | |
| BT005640.1 | - | 3 - 4 | 491 | 115 | ||
More Information
Set Membership: |
|

AT5G48650
Genomic Map