Assay Details
| Gene Symbol: AT5G48650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124241.4 | NP_199676.2 | 7 - 8 | 1279 | 82 | |
| GenBank mRNA | BT002864.1 | - | 7 - 8 | 1217 | 82 | |
| BT005640.1 | - | 6 - 7 | 1066 | 82 | ||
More Information
Set Membership: |
|

AT5G48650
Genomic Map