Assay Details
| Gene Symbol: PANC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124261.4 | NP_199695.1 | 1 - 2 | 880 | 137 | |
| GenBank mRNA | AK175238.1 | - | 1 - 2 | 593 | 137 | |
More Information
Set Membership: |
|

PANC
Genomic Map