Assay Details
| Gene Symbol: SAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124264.4 | NP_199698.1 | 1 - 2 | 97 | 109 | |
| GenBank mRNA | AY034896.1 | - | 1 - 2 | 36 | 109 | |
| BT025700.1 | - | 1 - 2 | 36 | 109 | ||
More Information
Set Membership: |
|

SAD1
Genomic Map