Assay Details
| Gene Symbol: GEX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344816.1 | NP_001330856.1 | 5 - 6 | 1291 | 62 | |
| NM_001344817.1 | NP_001330859.1 | 5 - 6 | 1291 | 62 | ||
| NM_001344818.1 | NP_001330860.1 | 5 - 6 | 1291 | 62 | ||
| NM_001344819.1 | NP_001330858.1 | 4 - 5 | 955 | 62 | ||
| NM_001344820.1 | NP_001330862.1 | 4 - 5 | 955 | 62 | ||
| NM_001344821.1 | NP_001330861.1 | 2 - 3 | 463 | 62 | ||
| NM_001344822.1 | NP_001330857.1 | 2 - 3 | 463 | 62 | ||
| NM_124292.3 | NP_199726.3 | 5 - 6 | 1262 | 62 | ||
| GenBank mRNA | AB743888.1 | - | 5 - 6 | 1380 | 62 | |
More Information
Set Membership: |
|

GEX2
Genomic Map