Assay Details
| Gene Symbol: MET1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344823.1 | NP_001331134.1 | 4 - 5 | 2844 | 72 | |
| NM_124293.4 | NP_199727.1 | 3 - 4 | 3153 | 72 | ||
| GenBank mRNA | L10692.1 | - | 3 - 4 | 3046 | 72 | |
More Information
Set Membership: |
|

MET1
Genomic Map