Assay Details
| Gene Symbol: MET1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344823.1 | NP_001331134.1 | 10 - 11 | 3941 | 93 | |
| NM_124293.4 | NP_199727.1 | 9 - 10 | 4250 | 93 | ||
| GenBank mRNA | L10692.1 | - | 9 - 10 | 4143 | 93 | |
More Information
Set Membership: |
|

MET1
Genomic Map