Assay Details
| Gene Symbol: SUS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344824.1 | NP_001331823.1 | 9 - 10 | 1485 | 66 | |
| NM_001344825.1 | NP_001331822.1 | 9 - 10 | 1397 | 66 | ||
| NM_124296.3 | NP_199730.1 | 10 - 11 | 1862 | 66 | ||
| GenBank mRNA | KJ138646.1 | - | 10 - 11 | 1463 | 66 | |
More Information
Set Membership: |
|

SUS2
Genomic Map