Assay Details
| Gene Symbol: SUS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344824.1 | NP_001331823.1 | 12 - 13 | 2277 | 66 | |
| NM_001344825.1 | NP_001331822.1 | 12 - 13 | 2189 | 66 | ||
| NM_124296.3 | NP_199730.1 | 13 - 14 | 2654 | 66 | ||
| GenBank mRNA | KJ138646.1 | - | 13 - 14 | 2255 | 66 | |
More Information
Set Membership: |
|

SUS2
Genomic Map