Assay Details
| Gene Symbol: SUS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344824.1 | NP_001331823.1 | 1 - 2 | 247 | 131 | |
| NM_001344825.1 | NP_001331822.1 | 1 - 2 | 159 | 131 | ||
| NM_124296.3 | NP_199730.1 | 2 - 3 | 624 | 131 | ||
| GenBank mRNA | KJ138646.1 | - | 2 - 3 | 225 | 131 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001344825.1 transcripts.
Set Membership: |
|

SUS2
Genomic Map