Assay Details
| Gene Symbol: PNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124335.3 | NP_199768.1 | 1 - 2 | 220 | 79 | |
| GenBank mRNA | AK228156.1 | - | 1 - 2 | 140 | 79 | |
| AM745095.1 | - | 1 - 2 | 76 | 79 | ||
| AY140065.1 | - | 1 - 2 | 136 | 79 | ||
| BT003398.1 | - | 1 - 2 | 76 | 79 | ||
More Information
Set Membership: |
|

PNG1
Genomic Map