Assay Details
| Gene Symbol: CLC-C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124367.4 | NP_199800.1 | 4 - 5 | 1135 | 72 | |
| GenBank mRNA | AY120754.1 | - | 4 - 5 | 966 | 72 | |
| BT001211.1 | - | 4 - 5 | 846 | 72 | ||
| Y09095.1 | - | 869 | 72 | |||
| Z71447.1 | - | 4 - 5 | 879 | 72 | ||
More Information
Set Membership: |
|

CLC-C
Genomic Map