Assay Details
| Gene Symbol: CLC-C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124367.4 | NP_199800.1 | 6 - 7 | 2543 | 96 | |
| GenBank mRNA | AY120754.1 | - | 6 - 7 | 2374 | 96 | |
| BT001211.1 | - | 6 - 7 | 2254 | 96 | ||
| Y09095.1 | - | 2277 | 96 | |||
| Z71447.1 | - | 6 - 7 | 2287 | 96 | ||
More Information
Set Membership: |
|

CLC-C
Genomic Map