Assay Details
| Gene Symbol: AT5G50100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124389.2 | NP_568719.1 | 1 - 2 | 239 | 79 | |
| GenBank mRNA | AY062766.1 | - | 1 - 2 | 168 | 79 | |
| AY081637.1 | - | 1 - 2 | 121 | 79 | ||
| AY087459.1 | - | 1 - 2 | 184 | 79 | ||
More Information
Set Membership: |
|

AT5G50100
Genomic Map