Assay Details
| Gene Symbol: AT5G50100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124389.2 | NP_568719.1 | 5 - 6 | 609 | 105 | |
| GenBank mRNA | AY062766.1 | - | 5 - 6 | 538 | 105 | |
| AY081637.1 | - | 5 - 6 | 491 | 105 | ||
| AY087459.1 | - | 5 - 6 | 554 | 105 | ||
More Information
Set Membership: |
|

AT5G50100
Genomic Map