Assay Details
| Gene Symbol: AT5G50170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124396.3 | NP_199828.1 | 1 - 2 | 558 | 67 | |
| GenBank mRNA | AY056116.1 | - | 1 - 2 | 436 | 67 | |
| BT009696.1 | - | 1 - 2 | 359 | 67 | ||
More Information
Set Membership: |
|

AT5G50170
Genomic Map