Assay Details
| Gene Symbol: UBC33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036971.2 | NP_001032048.2 | 5 - 6 | 449 | 123 | |
| NM_124425.4 | NP_199854.1 | 5 - 6 | 525 | 123 | ||
| GenBank mRNA | AF380654.1 | - | 5 - 6 | 467 | 123 | |
| AY142040.1 | - | 4 - 5 | 213 | 123 | ||
More Information
Set Membership: |
|

UBC33
Genomic Map