Assay Details
| Gene Symbol: UBC33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036971.2 | NP_001032048.2 | 8 - 9 | 571 | 100 | |
| NM_001036972.2 | NP_001032049.1 | 7 - 8 | 587 | 100 | ||
| NM_124425.4 | NP_199854.1 | 8 - 9 | 647 | 100 | ||
| GenBank mRNA | AF380654.1 | - | 8 - 9 | 589 | 100 | |
| AY142040.1 | - | 7 - 8 | 335 | 100 | ||
More Information
Set Membership: |
|

UBC33
Genomic Map