Assay Details
| Gene Symbol: UBC33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036971.2 | NP_001032048.2 | 9 - 10 | 677 | 81 | |
| NM_001036972.2 | NP_001032049.1 | 8 - 9 | 693 | 81 | ||
| NM_124425.4 | NP_199854.1 | 9 - 10 | 753 | 81 | ||
| GenBank mRNA | AF380654.1 | - | 9 - 10 | 695 | 81 | |
| AY142040.1 | - | 8 - 9 | 441 | 81 | ||
More Information
Set Membership: |
|

UBC33
Genomic Map