Assay Details
| Gene Symbol: TIM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124459.4 | NP_199894.1 | 1 - 2 | 251 | 66 | |
| GenBank mRNA | AF150083.1 | - | 1 - 2 | 125 | 66 | |
| BT002881.1 | - | 1 - 2 | 135 | 66 | ||
| BT004405.1 | - | 1 - 2 | 54 | 66 | ||
More Information
Set Membership: |
|

TIM8
Genomic Map