Assay Details
| Gene Symbol: EMB3142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344925.1 | NP_001318776.1 | 37 - 38 | 4627 | 79 | |
| NM_001344926.1 | NP_001332447.1 | 37 - 38 | 4910 | 79 | ||
| NM_001344927.1 | NP_001332446.1 | 37 - 38 | 4684 | 79 | ||
More Information
Set Membership: |
|

EMB3142
Genomic Map