Assay Details
| Gene Symbol: EMB3142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344925.1 | NP_001318776.1 | 40 - 41 | 4964 | 108 | |
| NM_001344926.1 | NP_001332447.1 | 40 - 41 | 5247 | 108 | ||
| NM_001344927.1 | NP_001332446.1 | 40 - 41 | 5018 | 108 | ||
More Information
Set Membership: |
|

EMB3142
Genomic Map