Assay Details
| Gene Symbol: ACD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344933.1 | NP_001331641.1 | 3 - 4 | 715 | 108 | |
| NM_124508.4 | NP_568756.2 | 3 - 4 | 715 | 108 | ||
| GenBank mRNA | AY362552.1 | - | 3 - 4 | 672 | 108 | |
More Information
Set Membership: |
|

ACD5
Genomic Map