Assay Details
| Gene Symbol: AT5G51300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124509.4 | NP_199943.1 | 1 | 2586 | 121 | |
| GenBank mRNA | AK317057.1 | - | 1 | 2531 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G51300| Gene Symbol: AT5G51300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124509.4 | NP_199943.1 | 1 | 2586 | 121 | |
| GenBank mRNA | AK317057.1 | - | 1 | 2531 | 121 | |
Set Membership: |
|