Assay Details
| Gene Symbol: AT5G51340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124513.3 | NP_199947.1 | 10 - 11 | 2238 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G51340| Gene Symbol: AT5G51340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124513.3 | NP_199947.1 | 10 - 11 | 2238 | 74 | |
Set Membership: |
|