Assay Details
| Gene Symbol: PLE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124539.4 | NP_199973.1 | 9 - 10 | 1703 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLE| Gene Symbol: PLE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124539.4 | NP_199973.1 | 9 - 10 | 1703 | 93 | |
Set Membership: |
|