Assay Details
| Gene Symbol: PGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124561.3 | NP_199995.1 | 4 - 5 | 794 | 75 | |
| GenBank mRNA | AF216580.1 | - | 4 - 5 | 509 | 75 | |
| AY099708.1 | - | 4 - 5 | 465 | 75 | ||
| AY128901.1 | - | 4 - 5 | 400 | 75 | ||
More Information
Set Membership: |
|

PGM
Genomic Map