Assay Details
| Gene Symbol: AT5G52390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124618.3 | NP_200052.1 | 2 - 3 | 686 | 85 | |
| GenBank mRNA | AY091116.1 | - | 2 - 3 | 490 | 85 | |
| AY122980.1 | - | 2 - 3 | 399 | 85 | ||
More Information
Set Membership: |
|

AT5G52390
Genomic Map