Assay Details
| Gene Symbol: AT5G53730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124752.4 | NP_200184.1 | 1 | 872 | 87 | |
| GenBank mRNA | BT010854.1 | - | 1 | 678 | 87 | |
| BT011320.1 | - | 1 | 581 | 87 | ||
More Information
Set Membership: |
|

AT5G53730
Genomic Map