Assay Details
| Gene Symbol: CUC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124774.3 | NP_200206.1 | 1 - 2 | 248 | 96 | |
| GenBank mRNA | BT030026.1 | - | 1 - 2 | 189 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUC2| Gene Symbol: CUC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124774.3 | NP_200206.1 | 1 - 2 | 248 | 96 | |
| GenBank mRNA | BT030026.1 | - | 1 - 2 | 189 | 96 | |
Set Membership: |
|