Assay Details
| Gene Symbol: AT5G53960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124775.2 | NP_200207.1 | 1 - 2 | 236 | 84 | |
| GenBank mRNA | DQ132753.1 | - | 1 - 2 | 236 | 84 | |
More Information
Set Membership: |
|

AT5G53960
Genomic Map