Assay Details
| Gene Symbol: HB52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124777.5 | NP_200209.1 | 1 | 693 | 78 | |
| GenBank mRNA | AK229211.1 | - | 1 | 531 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HB52| Gene Symbol: HB52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124777.5 | NP_200209.1 | 1 | 693 | 78 | |
| GenBank mRNA | AK229211.1 | - | 1 | 531 | 78 | |
Set Membership: |
|