Assay Details
| Gene Symbol: MOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336567.1 | NP_001325149.1 | 29 - 30 | 3629 | 85 | |
| NM_129117.5 | NP_565811.2 | 29 - 30 | 3323 | 85 | ||
| GenBank mRNA | AF367246.1 | - | 29 - 30 | 3069 | 85 | |
More Information
Set Membership: |
|

MOR1
Genomic Map