Assay Details
| Gene Symbol: ERF38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129125.2 | NP_181113.1 | 1 | 672 | 72 | |
| GenBank mRNA | AY560845.1 | - | 1 | 531 | 72 | |
| BT025754.1 | - | 1 | 531 | 72 | ||
| DQ446600.1 | - | 1 | 531 | 72 | ||
More Information
Set Membership: |
|

ERF38
Genomic Map