Assay Details
| Gene Symbol: NHL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129155.5 | NP_181140.1 | 1 | 660 | 55 | |
| GenBank mRNA | AK175879.1 | - | 1 | 524 | 55 | |
| AY085897.1 | - | 1 | 518 | 55 | ||
| BT025781.1 | - | 1 | 450 | 55 | ||
More Information
Set Membership: |
|

NHL12
Genomic Map