Assay Details
| Gene Symbol: YLS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129157.3 | NP_181142.1 | 1 | 900 | 136 | |
| GenBank mRNA | AB047812.1 | - | 1 | 786 | 136 | |
| AY086633.1 | - | 1 | 801 | 136 | ||
| BT002875.1 | - | 1 | 804 | 136 | ||
More Information
Set Membership: |
|

YLS9
Genomic Map