Assay Details
| Gene Symbol: TIM44-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129166.5 | NP_181151.3 | 13 - 14 | 1604 | 77 | |
| GenBank mRNA | BT015810.1 | - | 13 - 14 | 1400 | 77 | |
More Information
Set Membership: |
|

TIM44-2
Genomic Map