Assay Details
| Gene Symbol: CASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129169.3 | NP_181154.1 | 2 - 3 | 536 | 60 | |
| GenBank mRNA | AK228127.1 | - | 2 - 3 | 487 | 60 | |
| BT003113.1 | - | 2 - 3 | 406 | 60 | ||
More Information
Set Membership: |
|

CASP1
Genomic Map