Assay Details
| Gene Symbol: APG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129181.4 | NP_181165.1 | 1 - 2 | 204 | 69 | |
| GenBank mRNA | AB006139.1 | - | 1 - 2 | 184 | 69 | |
| AK118659.1 | - | 1 - 2 | 180 | 69 | ||
| AY140058.1 | - | 1 - 2 | 180 | 69 | ||
| BT002552.1 | - | 1 - 2 | 155 | 69 | ||
More Information
Set Membership: |
|

APG10
Genomic Map