Assay Details
| Gene Symbol: ABI5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336590.1 | NP_001324735.1 | 1 - 2 | 1222 | 89 | |
| NM_001336591.1 | NP_001324734.1 | 2 - 3 | 1416 | 89 | ||
| NM_129185.4 | NP_565840.1 | 2 - 3 | 1362 | 89 | ||
| GenBank mRNA | AF334206.1 | - | 2 - 3 | 1280 | 89 | |
| BT026517.1 | - | 1 - 2 | 1133 | 89 | ||
More Information
Set Membership: |
|

ABI5
Genomic Map