Assay Details
| Gene Symbol: LOS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336601.1 | NP_001324936.1 | 2 - 3 | 740 | 69 | |
| NM_129209.4 | NP_181192.1 | 2 - 3 | 541 | 69 | ||
| GenBank mRNA | AF424603.1 | - | 2 - 3 | 271 | 69 | |
| AY150418.1 | - | 2 - 3 | 198 | 69 | ||
More Information
Set Membership: |
|

LOS2
Genomic Map