Assay Details
| Gene Symbol: NAP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336618.1 | NP_001323430.1 | 11 - 12 | 2998 | 116 | |
| NM_129257.4 | NP_181238.4 | 11 - 12 | 2998 | 116 | ||
More Information
Set Membership: |
|

NAP12
Genomic Map