Assay Details
| Gene Symbol: RIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336624.1 | NP_001324128.1 | 3 - 4 | 1132 | 78 | |
| NM_129264.4 | NP_181245.1 | 3 - 4 | 607 | 78 | ||
| GenBank mRNA | BT020383.1 | - | 3 - 4 | 433 | 78 | |
| BT020462.1 | - | 2 - 3 | 196 | 78 | ||
More Information
Set Membership: |
|

RIP2
Genomic Map