Assay Details
| Gene Symbol: AT2G37200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336643.1 | NP_001325327.1 | 1 - 2 | 335 | 69 | |
| NM_129276.5 | NP_181257.2 | 1 - 2 | 279 | 69 | ||
| GenBank mRNA | AK228932.1 | - | 1 - 2 | 249 | 69 | |
| BT010678.1 | - | 1 - 2 | 272 | 69 | ||
| BT010966.1 | - | 1 - 2 | 222 | 69 | ||
More Information
Set Membership: |
|

AT2G37200
Genomic Map