Assay Details
| Gene Symbol: AT2G37200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336643.1 | NP_001325327.1 | 2 - 3 | 467 | 77 | |
| NM_129276.5 | NP_181257.2 | 2 - 3 | 411 | 77 | ||
| GenBank mRNA | AK228932.1 | - | 2 - 3 | 381 | 77 | |
| BT010678.1 | - | 2 - 3 | 404 | 77 | ||
| BT010966.1 | - | 2 - 3 | 354 | 77 | ||
More Information
Set Membership: |
|

AT2G37200
Genomic Map