Assay Details
| Gene Symbol: ABCG33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336645.1 | NP_001325069.1 | 11 - 12 | 2092 | 95 | |
| NM_001336646.1 | NP_001325068.1 | 10 - 11 | 2113 | 95 | ||
| NM_001336647.1 | NP_001325070.1 | 10 - 11 | 2094 | 95 | ||
| NM_129284.2 | NP_181265.1 | 11 - 12 | 2092 | 95 | ||
More Information
Set Membership: |
|

ABCG33
Genomic Map