Assay Details
| Gene Symbol: ABCG33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336645.1 | NP_001325069.1 | 17 - 18 | 3043 | 73 | |
| NM_001336646.1 | NP_001325068.1 | 16 - 17 | 3064 | 73 | ||
| NM_001336647.1 | NP_001325070.1 | 16 - 17 | 3045 | 73 | ||
| NM_129284.2 | NP_181265.1 | 17 - 18 | 3043 | 73 | ||
More Information
Set Membership: |
|

ABCG33
Genomic Map